missing translation for 'onlineSavingsMsg'
Learn More

AF9 Antibody [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30510082 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30510082 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510082 Supplier Novus Biologicals Supplier No. NBP338336G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AF9 Polyclonal antibody specifically detects AF9 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AF9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias AF-9, AF9myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 3, ALL1-fused gene from chromosome 9 protein, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 3, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 3, Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein, protein AF-9, YEATS domain-containing protein 3, YEATS3FLJ2035
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human AF9 (NP_004520.2).,, Sequence:, MASSCAVQVKLELGHRAQVRKKPTVEGFTHDWMVFVRGPEHSNIQHFVEKVVFHLHESFPRPKRVCKDPPYKVEESGYAGFILPIEVYFKNKEEPRKVRFDYDLFLHLEGHPPVNHLRCEKLTFNNPTEDFRRKLLKAGGDPNRSIHTSS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4300
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.