missing translation for 'onlineSavingsMsg'
Learn More

AG-3/AGR3 Antibody (AGR3.1), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18449543 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18449543 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18449543 Supplier Novus Biologicals Supplier No. NBP145004AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AG-3/AGR3 Monoclonal antibody specifically detects AG-3/AGR3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AG-3/AGR3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone AGR3.1
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias AG-3, anterior gradient homolog 3 (Xenopus laevis), BCMP11AG3, Breast cancer membrane protein 11, hAG-3member 18
Host Species Mouse
Immunogen Purified human AGR3 protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 155465
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.