missing translation for 'onlineSavingsMsg'
Learn More

AGER Antibody (CL10587) [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30530065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30530065 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30530065 Supplier Novus Biologicals Supplier No. NBP344242V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AGER Monoclonal antibody specifically detects AGER in Human,Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGER
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL10587
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q15109, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 177
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.