missing translation for 'onlineSavingsMsg'
Learn More

AGFG2 Antibody - BSA Free, Novus Biologicals™

Product Code. 30558891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558891 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558891 Supplier Novus Biologicals Supplier No. NBP348201

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AGFG2 Polyclonal antibody specifically detects AGFG2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen AGFG2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA, Immunohistochemistry 1:20-1:200, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias ArfGAP with FG repeats 2, HIV-1 Rev binding protein-like, HIV-1 Rev-binding protein-like protein, HRBLnucleoporin, RAB-R, RABRarf-GAP domain and FG repeats-containing protein 2, Rev/Rex activation domain binding protein-related, Rev/Rex activation domain-binding protein related
Host Species Rabbit
Immunogen ArfGAP with FG repeats 2. (Uniprot# O95081)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3268
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.