missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIP/ARA9 Antibody (35-2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-127
This item is not returnable.
View return policy
Description
AIP/ARA9 Monoclonal specifically detects AIP/ARA9 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| AIP/ARA9 | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.1% Sodium Azide | |
| ARA9, FKBP16, FKBP37, SMTPHN, XAP2, XAP-2 | |
| Mouse | |
| 37 kDa | |
| 0.1 mL | |
| Apoptosis, Hypoxia, Neuroscience, Neurotransmission | |
| 9049 | |
| Human, Mouse, Rat | |
| Supernatant |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 35-2 | |
| Western Blot 1:1000, Simple Western 1:200, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23702468), Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 21984905) | |
| O00170 | |
| AIP | |
| Bacterially expressed human AIP/ARA9 [UniProt# O00170] | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Customers have reported success on rat lysate (see review). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction