missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIP/ARA9 Antibody (35-2), Novus Biologicals™
Mouse Monoclonal Antibody
225.00 € - 533.00 €
Specifications
| Antigen | AIP/ARA9 |
|---|---|
| Clone | 35-2 |
| Dilution | Western Blot 1:1000, Simple Western 1:200, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23702468), Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 21984905) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057495
|
Novus Biologicals
NB100-127SS |
0.025 mL |
225.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717613
|
Novus Biologicals
NB100-127 |
0.1 mL |
533.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIP/ARA9 Monoclonal specifically detects AIP/ARA9 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| AIP/ARA9 | |
| Western Blot 1:1000, Simple Western 1:200, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23702468), Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 21984905) | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human, Mouse, Rat | |
| O00170 | |
| 9049 | |
| Bacterially expressed human AIP/ARA9 [UniProt# O00170] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 37 kDa |
| 35-2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Hypoxia, Neuroscience, Neurotransmission | |
| Tissue culture supernatant with 0.1% Sodium Azide | |
| ARA9, FKBP16, FKBP37, SMTPHN, XAP2, XAP-2 | |
| AIP | |
| IgG1 | |
| Tissue culture supernatant | |
| Customers have reported success on rat lysate (see review). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title