missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK3L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47553-25ul
This item is not returnable.
View return policy
Description
AK3L1 Polyclonal specifically detects AK3L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AK3L1 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AK4 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PEAVAARLRQYKDVAKPVIELYKSRGVLHQF | |
| 25 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 205 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction