missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AKAP4 Polyclonal antibody specifically detects AKAP4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | AKAP4 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | A kinase (PRKA) anchor protein 4, AKAP-4, AKAP82AKAP 82, A-kinase anchor protein 4, A-kinase anchor protein 82 kDa, cancer/testis antigen 99, CT99, hAKAP82Fsc1, HIFSC1, Major sperm fibrous sheath protein, p82, PRKA4, protein kinase A anchoring protein 4, Protein kinase A-anchoring protein 4, testis-specific gene HI |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 189-340 of human AKAP4 (NP_003877.2).,, Sequence:, QSPSAPPAKPPSTQRAVISPDGECSIDDLSFYVNRLSSLVIQMAHKEIKEKLEGKSKCLHHSICPSPGNKERISPRTPASKIASEMAYEAVELTAAEMRGTGEESREGGQKSFLYSELSNKSKSGDKQMSQRESKEFADSISKGLMVYANQV |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?