missing translation for 'onlineSavingsMsg'
Learn More

Albumin Antibody Pair, Novus Biologicals™

Product Code. 18321248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321248 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321248 Supplier Novus Biologicals Supplier No. H00000213AP11
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Pair

Albumin antibody specifically detects Albumin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Albumin
Applications Sandwich ELISA
Conjugate Unconjugated
Dilution Sandwich ELISA 10 ng/mL - 100 ng/mL
Formulation 1x PBS, pH 7.4
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Quantity 1 Set
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.