Learn More
Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (23GB4315), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552683
 View more versions of this product
Product Code. 30087822
  • 186.00 € / 20µL
Description
This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ALDH16A1
Recombinant Monoclonal
Unconjugated
ALDH16A1
2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209
Rabbit
Affinity chromatography
RUO
126133, 361571, 69748
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB4315
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q3T1L0, Q571I9, Q8IZ83
ALDH16A1
A synthesized peptide derived from human ALDH16A1 (550-590AA).
20 μL
Primary
Human, Mouse, Rat