missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (23GB4315), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | ALDH16A1 |
|---|---|
| Clone | 23GB4315 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089015
|
Invitrogen™
MA552682 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087822
|
Invitrogen™
MA552683 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH16A1 Recombinant Monoclonal antibody specifically detects ALDH16A1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ALDH16A1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q3T1L0, Q571I9, Q8IZ83 | |
| 126133, 361571, 69748 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4315 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209 | |
| A synthesized peptide derived from human ALDH16A1 (550-590AA). | |
| Primary | |
| -20°C | |
| ALDH16A1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title