missing translation for 'onlineSavingsMsg'
Learn More

Aldolase C Antibody (6F3) [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30518233 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30518233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518233 Supplier Novus Biologicals Supplier No. NBP342925AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aldolase C Monoclonal antibody specifically detects Aldolase C in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase C
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 6F3
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C
Host Species Mouse
Immunogen aldolase C, fructose-bisphosphate - (Uniprot# P09972)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 230
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.