missing translation for 'onlineSavingsMsg'
Learn More

ALK/CD246 Mouse anti-Human, Clone: ALK/1032, R&D Systems™

Product Code. 18609538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609538 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609538 Supplier Novus Biologicals Supplier No. NBP307749100ug
  • ONLINE EXCLUSIVE PRICE
    566.80 €
    479.85 € / 100µg
    Save 86.95 €
    15% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALK/CD246
Applications Western Blot
Classification Monoclonal
Clone ALK/1032
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Host Species Mouse
Immunogen Recombinant fragment (around aa 220-335) of human ALK/CD246 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 238
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.