missing translation for 'onlineSavingsMsg'
Learn More

ALKBH8 Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30511663 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511663 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511663 Supplier Novus Biologicals Supplier No. NBP338393AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ALKBH8 Polyclonal antibody specifically detects ALKBH8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH8
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ABH8MGC10235, AlkB homologue 8, alkB, alkylation repair homolog 8 (E. coli), alkylated DNA repair protein alkB homolog 8, EC 1.14.11.-, EC 2.1.1.-, FLJ38204, Probable alpha-ketoglutarate-dependent dioxygenase ABH8, S-adenosyl-L-methionine-dependent tRNA methyltransferase ABH8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human ALKBH8 (NP_620130.2).,, Sequence:, MDSNHQSNYKLSKTEKKFLRKQIKAKHTLLRHEGIETVSYATQSLVVANGGLGNGVSRNQLLPVLEKCGLVDALLMPPNKPYSFARYRTTEESKRAYVTLNGKEVVDDLGQKITLYLNFVEKVQWKELRPQALPPGLMVVEEIISSEEEKMLLESVDWTEDTDNQNSQKSLKHRRVKHFGYEFHYENNNVDKDKPLSGGLPDICESFLEKWLRKGYIKHKPDQMTINQYE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91801
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.