missing translation for 'onlineSavingsMsg'
Learn More

all-rac-alpha-Tocopherol (Vitamin E), Mikromol™

Product Code. 17951616
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951616 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951616 Supplier Mikromol Supplier No. MM0167.000250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

all-rac-alpha-Tocopherol (Vitamin E)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-alpha-Tocopherol
CAS 10191-41-0
Analyte or Component Names DL-alpha-Tocopherol
Shipping Condition Ice pack (-20°C)
Molecular Formula C29 H50 O2
Certifications/Compliance ISO 17034
Synonym 6-Chromanol, 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)- (8CI), 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-1-benzopyran-6-ol, (±)-α-Tocopherol, 2,5,7,8-Tetramethyl-2-(4',8',12'-trimethyltridecyl)chroman-6-ol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, DL-α-Tocopherol, Elementol B, Elementol Basic, Elementol R, Ephanyl, Irganox E 210, Rac-α-Tocopherol, Ronotec 201, Ronotec 202, Ronotec DF 120, Uvinul 2000AO, all-rac-α-Tocopherol, dl-α-Tocopherol
InChI Formula InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol
Molecular Weight (g/mol) 430.71
Quantity 250 mg
Formula Weight 430.3811 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.