missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha COP I Antibody (SR1142), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22116-100ul
This item is not returnable.
View return policy
Description
alpha COP I Monoclonal antibody specifically detects alpha COP I in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| alpha COP I | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1142 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
| alpha coat protein, Alpha-coat protein, Alpha-COP, coatomer protein complex, subunit alpha, coatomer subunit alpha, FLJ26320, HEPCOP, HEP-COPalpha-COP, xenin | |
| A synthesized peptide derived from human alpha COP I (Uniprot #: P53621) | |
| 100 μg | |
| Signal Transduction | |
| 1314 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction