missing translation for 'onlineSavingsMsg'
Learn More

alpha COP I Antibody (SR1142), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00 € - 449.00 €

Specifications

Antigen alpha COP I
Clone SR1142
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18601484
View Documents
Novus Biologicals
NBP3-22116-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18690864
View Documents
Novus Biologicals
NBP3-22116-50ul
50 μg
338.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha COP I Monoclonal antibody specifically detects alpha COP I in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

alpha COP I
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
alpha coat protein, Alpha-coat protein, Alpha-COP, coatomer protein complex, subunit alpha, coatomer subunit alpha, FLJ26320, HEPCOP, HEP-COPalpha-COP, xenin
A synthesized peptide derived from human alpha COP I (Uniprot #: P53621)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1142
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
1314
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.