missing translation for 'onlineSavingsMsg'
Learn More

alpha-Resorcylic Acid (3,5-Dihydroxybenzoic Acid), LoGiCal

Product Code. 30463092
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463092 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463092 Supplier LoGiCal Supplier No. LGCFOR0708.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Resorcylic acid
CAS 99-10-5
Analyte or Component Names alpha-Resorcylic acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O4
Certifications/Compliance ISO 9001
Synonym Terbutaline Sulphate Imp. A (EP), Terbutaline Imp. A (EP), 3,5-Dihydroxybenzoic Acid, alpha-Resorcylic Acid, alpha-Resorcylic acid, 3,5-Dihydroxybenzoic acid, 5-Carboxyresorcinol, NSC 22948, Terbutaline Sulfate Impurity A, Terbutaline Impurity A
InChI Formula InChI=1S/C7H6O4/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,8-9H,(H,10,11)
Solution Type Neat
SMILES OC(=O)c1cc(O)cc(O)c1
IUPAC Name 3,5-dihydroxybenzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 154.12
Quantity 10 mg
Formula Weight 154.0266 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.