missing translation for 'onlineSavingsMsg'
Learn More

alpha-Smooth Muscle Actin Antibody Pair, Novus Biologicals™

Product Code. 18390168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18390168 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390168 Supplier Novus Biologicals Supplier No. H00000059AP41
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Pair

alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Smooth Muscle Actin
Applications Sandwich ELISA
Classification Monoclonal
Conjugate Unconjugated
Dilution Sandwich ELISA 0.3 to 100 ng/mL
Formulation 1x PBS, pH 7.4
Gene Alias AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Quantity 1 Set
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.