missing translation for 'onlineSavingsMsg'
Learn More

alpha-Synuclein, p Tyr136 Antibody - Azide and BSA Free, R&D Systems™

Product Code. 18612429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18612429 20 μg 20µL
18638448 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612429 Supplier Novus Biologicals Supplier No. NBP30540420ul
 Restricted Item
Estimated Shipment: 02-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

alpha-Synuclein Polyclonal specifically detects alpha-Synuclein in Human/Mouse/Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen alpha-Synuclein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y136 of human alpha-Synuclein (NP_000336.1). QDYEP
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6622
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.