missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amfenac Methyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Amfenac Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Amfenac Methyl Ester |
| CAS | 61941-58-0 |
| Analyte or Component Names | Amfenac Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Amfenac Methyl Ester, 2-Amino-3-benzoylbenzeneacetic acid methyl ester, Methyl (2-amino-3-benzoylphenyl)acetate, Methyl 2-amino-3-benzoylbenzeneacetate |
| InChI Formula | InChI=1S/C16H15NO3/c1-20-14(18)10-12-8-5-9-13(15(12)17)16(19)11-6-3-2-4-7-11/h2-9H,10,17H2,1H3 |
| SMILES | COC(=O)Cc1cccc(C(=O)c2ccccc2)c1N |
| IUPAC Name | methyl 2-(2-amino-3-benzoylphenyl)acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction