missing translation for 'onlineSavingsMsg'
Learn More

Amifampridin, Mikromol™

Product Code. 30050504
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050504 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050504 Supplier Mikromol Supplier No. MM3483.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amifampridin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amifampridin
CAS 54-96-6
Analyte or Component Names Amifampridin
Shipping Condition Room Temperature
Molecular Formula C5 H7 N3
Certifications/Compliance ISO 17025
Synonym 3,4-Pyridinediamine, Pyridine, 3,4-diamino- (6CI,7CI,8CI), 3,4-DAP, 3,4-Diaminopyridine, 4,5-Diaminopyridine, Amifampridine, DAP, NSC 521760, Pyridine-4,5-diamine
InChI Formula InChI=1S/C5H7N3/c6-4-1-2-8-3-5(4)7/h1-3H,7H2,(H2,6,8)
SMILES Nc1ccncc1N
IUPAC Name pyridine-3,4-diamine
Molecular Weight (g/mol) 109.13
Quantity 100 mg
Formula Weight 109.064 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.