missing translation for 'onlineSavingsMsg'
Learn More

AMIGO3 Antibody (CL11023) [Allophycocyanin], Novus Biologicals™

Product Code. 30297188 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
30297188 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy

Product Code. 30297188

Brand: Novus Biologicals NBP307991APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AMIGO3 Monoclonal antibody specifically detects AMIGO3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMIGO3
Applications Western Blot
Classification Monoclonal
Clone CL11023
Conjugate APC
Formulation PBS
Gene Alias adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from Q86WK7, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386724
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.