missing translation for 'onlineSavingsMsg'
Learn More

Amikacin (From Kanamycin A, White Crystalline Powder), Fisher BioReagents

Product Code. p-24173
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10134924 250 mg Amber Glass 250mg
10011313 1 g Amber Glass 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10134924 Supplier Fisher BioReagents Supplier No. BP2643250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • An antibiotic that serves as a selective agent to isolate transformed bacteria
  • Amikacin functions as an indirect inhibitor of protein synthesis in prokaryotes by binding to the 30S subunit of ribosomes
  • Useful as a selective agent to isolate bacteria that have been transformed with plasmid containing the antibiotic-resistant gene
  • Intended for research purposes only; not for manufacturing or commercial use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-28-5
Molecular Formula C22H43N5O13
Molecular Weight (g/mol) 585.608
InChI Key LKCWBDHBTVXHDL-RMDFUYIESA-N
Synonym Lukadin
PubChem CID 37768
ChEBI CHEBI:2637
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N

Specifications

Type Amikacin
Physical Form Solid
pH 9.5 to 11.5
Melting Point 214°C
Packaging Amber Glass
Potency > 900μg/mg
Color White
Quantity 250 mg
Merck Index 15, 400
Solubility Information completely soluble in water
Formula Weight 585.61
Chemical Name or Material Amikacin, From Kanamycin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.