accessibility menu, dialog, popup

UserName

CAS RN 37517-28-5

IUPAC Name: (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide
Synonyms: amikacin amikason's son's brand of amikacin sulfate amikacin dihydrate galen brand of amikacin sulfate amukin amikacine amikacina son brand of amikacin sulfate biklin
Molecular Weight (g/mol): 585.61
Molecular Formula: C22H43N5O13
InChi Key: LKCWBDHBTVXHDL-RMDFUYIESA-N
SMILES: NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Color 
  • (2)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)