missing translation for 'onlineSavingsMsg'
Learn More

Aminocaproic Acid, Mikromol™

Product Code. 17909404
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17909404 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17909404 Supplier Mikromol Supplier No. MM0896.00
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aminocaproic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminocaproic acid
CAS 60-32-2
Analyte or Component Names Aminocaproic acid
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O2
Certifications/Compliance ISO 17034
Synonym Aminocaproic Acid, Ω-Aminocaproic acid, Ω-Aminohexanoic acid, 177 J.D., 6-Amino-n-hexanoic acid, 6-Aminocaproic acid, 6-Aminohexanoic acid, ACS, Acepramin, Acepramine, Acikaprin, Afibrin, Amicar, Amikar, Aminokapron, CL 10304, CY 116, Caplamin, Capramol, Caprocid, Caprolisin, EACA, EACS, Epsamon, Epsicapron, Epsikapron, Epsilcapramin, Epsilon S, Hemocaprol, Hemopar, Hepin, Ipsilon, NSC 212532, NSC 26154, NSC 400230, Respramin, epsilon-Aminocaproic acid, epsilon-Amino-n-hexanoic acid, epsilon-Aminohexanoic acid, epsilon-Leucine, epsilon-Norleucine, omega-Aminocaproic acid, omega-Aminohexanoic acid
InChI Formula InChI=1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)
SMILES NCCCCCC(=O)O
IUPAC Name 6-aminohexanoic acid
Molecular Weight (g/mol) 131.17
Quantity 500 mg
Formula Weight 131.0946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.