missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aminopeptidase P1/XPNPEP1 Polyclonal specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Western Blot.
Specifications
Specifications
| Antigen | Aminopeptidase P1/XPNPEP1 |
| Applications | Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Aminopeptidase P1/XPNPEP1 fragment. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Política de privacidad.
¿Detecta una oportunidad de mejora?