missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aminopeptidase P2/XPNPEP2 Polyclonal antibody specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Aminopeptidase P2/XPNPEP2 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Aminopeptidase P2/XPNPEP2 (Accession#: O43895; Met 1-Ala 650) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?