missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline.HCl, Lipomed

Product Code. 30462309
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462309 50 mg 50mg
30462332 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30462309 Supplier Lipomed Supplier No. LPMAMI754HC50
 Restricted Item
Estimated Shipment: 04-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Amitriptyline.HCl
TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Amitriptyline Hydrochloride
CAS 549-18-8
Analyte or Component Names Amitriptyline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H23 N . Cl H
Synonym Amitriptyline Hydrochloride, Nortriptyline Hydrochloride Imp. F (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (9 CI), 5-(3-Dimethylaminopropylidene)dibenzo[a,d][1,4 ]cycloheptadiene hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, 10,11-Dihydro-N,N-dimethyl-5 H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CN(C)CCC=C1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7 ]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 313.86
Quantity 50 mg
Formula Weight 313.1597 g/mol
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.