missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314435
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314435 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314435 Supplier Dr. Ehrenstorfer Supplier No. DREA10231000AL100
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amitriptyline Hydrochloride
CAS 549-18-8
Analyte or Component Names Amitriptyline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H23 N . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Amitriptyline Hydrochloride, Nortriptyline Hydrochloride Imp. F (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (9CI), 5-(3-Dimethylaminopropylidene)dibenzo[a,d][1,4]cycloheptadiene hydrochloride, ADT, Adepril, Amicen, Amil 25, Amilent, Amilit, Amineurin, Amiplin, Amiprin, Amitid, Amitril, Amitrip, Amitriptyline chloride, Amyline, Amyzol, Anapsique, Apo-Amitriptyline, Damilen hydrochloride, Deprex, Domical, Elatrol, Elatrolet, Elavil, Enafon, Endep, Etravil, Euplit, Lantron, Laroxyl, Larozyl, Lentizol, Miketorin, NIH 10794, Noriline, Novoprotect, Pinsanu, Pinsaun, Redomex, Ro 4-1575, Saroten, Saroten Retard, Sarotena, Sarotex, Sylvemid, Syneudon, Teperin, Trepiline, Tridep, Tripta, Triptizol, Trynol, Tryptal, Tryptanol, Tryptine, Tryptizol, Trytomer, Uxen, Vanatrip, 5H-Dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine, 10,11-dihydro-N,N-dimethyl-, hydrochloride (6CI,8CI), 10,11-Dihydro-N,N-dimethyl-5H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, Nortriptyline Hydrochloride Impurity F as Hydrochloride, Nortriptyline Impurity F as Hydrochloride
InChI Formula InChI=1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H
SMILES Cl.CN(C)CCC=C1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 313.86
Quantity 1 mL
Formula Weight 313.1597 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.