missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amodiaquine dihydrochloride dihydrate

Product Code. p-7077535 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15314998

Brand: Thermo Scientific Chemicals 455200050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6398-98-7
Molecular Formula 2HCl·2H2O
Molecular Weight (g/mol) 464.81
InChI Key YVNAYSHNIILOJS-UHFFFAOYSA-N
Synonym amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1
PubChem CID 64646
ChEBI CHEBI:50652
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
SMILES CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl

Specifications

Melting Point 166.0°C to 172.0°C
Boiling Point 478.0°C
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 464.81
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Amodiaquine dihydrochloride dihydrate
compliance-icons
Product Identifier
  • Amodiaquine dihydrochloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.