accessibility menu, dialog, popup

UserName

HHH2OH2OClClH3CNH3CNHNClHO

CAS RN 6398-98-7

IUPAC Name: dihydrogen 4-[(7-chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrate dichloride
Synonyms: flavoquine amodiaquin dihydrochloride dihydrate
Molecular Weight (g/mol): 464.81
Molecular Formula: C20H28Cl3N3O3
InChi Key: YVNAYSHNIILOJS-UHFFFAOYSA-N
SMILES: [H+].[H+].O.O.[Cl-].[Cl-].CCN(CC)CC1=CC(NC2=C3C=CC(Cl)=CC3=NC=C2)=CC=C1O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)