missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Trihydrate, LoGiCal

Product Code. 30459336
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459336 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459336 Supplier LoGiCal Supplier No. LGCFOR0232.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amoxicillin Trihydrate
CAS 61336-70-7
Analyte or Component Names Amoxicillin Trihydrate
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H19 N3 O5 S . 3 H2 O
Certifications/Compliance ISO 9001
Synonym Amoxicillin Trihydrate, (2S,5R,6R)-6-[[(2R)-Amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, [2S-[2alpha,5alpha,6beta(S*)]]-6-[[Amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxycillin trihydrate, (-)-6-[2-amino-2-(p-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0] heptane-2-carboxylic acid, 6-[D(-)-alpha-amino-p-hydroxyphenylacetamido]penicillanic acid, alpha-amino-p-hydroxybenzylpenicillin, 6-(p-hydroxy-alpha-aminophenylacetamido)penicillanic acid, p-hydroxyampicillin, amoxycillin, BRL-2333, Amoxicillin trihydrate
InChI Formula InChI=1S/C16H19N3O5S.3H2O/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;;;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);3*1H2/t9-,10-,11+,14-;;;/m1.../s1
Solution Type Neat
SMILES O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
Industry Type Clinical Testing
Molecular Weight (g/mol) 419.45
Quantity 10 mg
Formula Weight 419.1362 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.