missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Trihydrate, Mikromol™

Product Code. 17974026
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974026 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17974026 Supplier Mikromol Supplier No. MM0232.000250
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amoxicillin Trihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amoxicillin Trihydrate
CAS 61336-70-7
Analyte or Component Names Amoxicillin Trihydrate
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H19 N3 O5 S . 3 H2 O
Certifications/Compliance ISO 17034
Synonym Amoxicillin Trihydrate, (2S,5R,6R)-6-[[(2R)-Amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, [2S-[2alpha,5alpha,6beta(S*)]]-6-[[Amino(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxycillin trihydrate, (-)-6-[2-amino-2-(p-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0] heptane-2-carboxylic acid, 6-[D(-)-alpha-amino-p-hydroxyphenylacetamido]penicillanic acid, alpha-amino-p-hydroxybenzylpenicillin, 6-(p-hydroxy-alpha-aminophenylacetamido)penicillanic acid, p-hydroxyampicillin, amoxycillin, BRL-2333, Amoxicillin trihydrate
InChI Formula InChI=1S/C16H19N3O5S.3H2O/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;;;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);3*1H2/t9-,10-,11+,14-;;;/m1.../s1
SMILES O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
Molecular Weight (g/mol) 419.45
Quantity 250 mg
Formula Weight 419.1362 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.