missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin, anhydrous, Mikromol™

Product Code. 17980896
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980896 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980896 Supplier Mikromol Supplier No. MM0253.16
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ampicillin, anhydrous
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ampicillin
CAS 69-53-4
Analyte or Component Names Ampicillin
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H19 N3 O4 S
Certifications/Compliance ISO 17034
Synonym Ampicillin, anhydrous, (2S,5R,6R)-6-[[(2R)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, CCI 5478, BRL-1341, Sultamicillin Impurity C, Sultamicillin Tosilate Dihydrate Impurity B, Sultamicillin Tosilate Impurity B
InChI Formula InChI=1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23)/t9-,10-,11+,14-/m1/s1
SMILES CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)O
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 349.4
Quantity 500 mg
Formula Weight 349.1096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.