missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (S06-1F9), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | Angiopoietin-1 |
|---|---|
| Clone | S06-1F9 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:1000, RNA Inhibition |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673695
|
Novus Biologicals
NBP3-19492-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647064
|
Novus Biologicals
NBP3-19492-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-1 Monoclonal antibody specifically detects Angiopoietin-1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, RNA InhibitionSpecifications
| Angiopoietin-1 | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 284 | |
| IgG | |
| Affinity purified |
| S06-1F9 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:1000, RNA Inhibition | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| A synthetic peptide of human Angiopoietin-1 (Uniprot # Q15389) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title