missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrochloride, Mikromol™

Product Code. 18030940
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18030940 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18030940 Supplier Mikromol Supplier No. MM0093.17
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aniline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aniline Hydrochloride
CAS 142-04-1
Analyte or Component Names Aniline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H7 N . Cl H
Certifications/Compliance ISO 17025
Synonym 4-Aminobenzoic Acid Imp. C (EP), Fentanyl Imp. F (EP), Mesalazine Imp. K (EP), Sodium Cyclamate Imp. B (EP), Trimethoprim Imp. K (EP), Aniline, Aniline Hydrochloride, Phenylamine Hydrochloride, 4-Aminobenzoic Acid Imp. C (EP) as Hydrochloride, Sodium Cyclamate Imp. B (EP) as Hydrochloride, Fentanyl Imp. F (EP) as Hydrochloride, Mesalazine Imp. K (EP) as Hydrochloride, Trimethoprim Imp. K (EP) as Hydrochloride, 4-Aminobenzoic Acid Impurity C as Hydrochloride, Fentanyl Impurity F as Hydrochloride, Mesalazine Impurity K as Hydrochloride, Sodium Cyclamate Impurity B as Hydrochloride, Trimethoprim Impurity K as Hydrochloride
InChI Formula InChI=1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H
SMILES Cl.Nc1ccccc1
IUPAC Name aniline;hydrochloride
Molecular Weight (g/mol) 129.59
Quantity 100 mg
Formula Weight 129.0345 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.