missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKMY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00 € - 326.00 €
Specifications
| Antigen | ANKMY1 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639741
|
Novus Biologicals
NBP2-97343-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632514
|
Novus Biologicals
NBP2-97343-50ul |
50 μg |
241.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANKMY1 Polyclonal antibody specifically detects ANKMY1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| ANKMY1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ankyrin repeat and MYND domain containing 1, DKFZp686D20155, TSAL1, ZMYND13 | |
| Produced in rabbits immunized with E. coli-derived Human ANKMY1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 51281 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title