missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83532-25ul
This item is not returnable.
View return policy
Description
ANKS3 Polyclonal antibody specifically detects ANKS3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ANKS3 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| ankyrin repeat and SAM domain-containing protein 3, ankyrin repeat and sterile alpha motif domain containing 3, FLJ32767, KIAA1977FLJ32345 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LNHGVKVDARDHSGATARMLAKQYGHMKIVALMDTYSPSLPKSLYRSPEKYEDLSSSDESCPAPQRQRPCRKKGVSI | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 124401 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction