missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 624.00 €
Specifications
| Antigen | ANKS3 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18400041
|
Novus Biologicals
NBP1-83532 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18490141
|
Novus Biologicals
NBP1-83532-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANKS3 Polyclonal antibody specifically detects ANKS3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ANKS3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ankyrin repeat and SAM domain-containing protein 3, ankyrin repeat and sterile alpha motif domain containing 3, FLJ32767, KIAA1977FLJ32345 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LNHGVKVDARDHSGATARMLAKQYGHMKIVALMDTYSPSLPKSLYRSPEKYEDLSSSDESCPAPQRQRPCRKKGVSI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 124401 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title