missing translation for 'onlineSavingsMsg'
Learn More

Annexin A2 Antibody (10H9), Novus Biologicals™

Product Code. p-200022270 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
50 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30043450 50 μL 50µL
30043758 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30043450 Supplier Novus Biologicals Supplier No. NBP32650650ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Annexin A2 Monoclonal antibody specifically detects Annexin A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 10H9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Annexin A2 [UniProt P07355]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 302
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.