missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%

Product Code. 15459415
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459415 1 g 1g
15469415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459415 Supplier Thermo Scientific Alfa Aesar Supplier No. H51921.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 863992-56-7 | C26H32B2O4 | 430.16 g/mol

It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n′-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863992-56-7
Molecular Formula C26H32B2O4
Molecular Weight (g/mol) 430.16
MDL Number MFCD16294538
InChI Key ZLXSWNVYGSZXOP-UHFFFAOYSA-N
Synonym 9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl
PubChem CID 57415691
IUPAC Name 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12

Specifications

Quantity 1 g
Formula Weight 430.16
Percent Purity 95%
Chemical Name or Material Anthracene-9,10-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.