missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9-carboxylic acid, 98+%

Product Code. 11467956
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467956 250 g 250g
11447956 10 g 10g
11457956 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467956 Supplier Thermo Scientific Alfa Aesar Supplier No. A14049.30
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 723-62-6 | C15H10O2 | 222.24 g/mol

Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723-62-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00001257
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
PubChem CID 2201
ChEBI CHEBI:34507
IUPAC Name anthracene-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point ∼218°C (decomposition)
Quantity 250 g
Beilstein 1875336
Solubility Information Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene,chloroform,and hydronaphthalenes. Insoluble in water.
Formula Weight 222.24
Percent Purity ≥98%
Chemical Name or Material Anthracene-9-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.