accessibility menu, dialog, popup

UserName

HOO

CAS RN 723-62-6

IUPAC Name: anthracene-9-carboxylic acid
Synonyms: ANCA 9-aca 9-AC cpd sodium salt 9-anthroic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: XGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)