Learn More
Btk Mouse anti-Human, Unlabeled, Clone: 53/BTK, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611117
Description
Bruton's tyrosine kinase (Btk) is a nonreceptor tyrosine kinase whose function is critical for proper B cell development and signaling. It is a member of the Tec family of kinases which includes Tec and Itk. In addition to an N-terminal pleckstrin homology (PH) domain, the Tec proteins contain Src homology domains 2 and 3 (SH2 and SH3) and a stretch of 60-80 amino acids between the PH and SH3 domains termed the Tec homology domain. The activity of Btk is regulated by Src-mediated phosphorylation of the kinase domain at tyrosine 551. This event induces Btk kinase activity and subsequent autophosphorylation at tyrosine 223 in the SH3 domain. Phosphorylated Btk then associates with the cell membrane via the interaction of the PH domain with phosphatidylinositol 3, 4, 5-triphosphate. The PH domain is essential for proper activation and function of Btk. A mutation in the PH domain results in Xid, murine X-linked immunodeficiency, and human X-linked agammaglobulinemia.
Host Species: Mouse
Clone: 53
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human N-Terminal Btk aa. 2-172 Recombinant Protein
Formula Weight [Chemical]: 77kDa
Immunofluorescence, Western Blotting
Specifications
| Btk | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human N-Terminal Btk aa. 2-172 Recombinant Protein | |
| 150 μg | |
| Cell Biology | |
| Human | |
| IgG2a |
| Western Blot | |
| 53/BTK | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.