missing translation for 'onlineSavingsMsg'
Learn More

CD178 Mouse anti-Human, Biotin, Clone: NOK-1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 556373

 View more versions of this product

Product Code. 15818678

  • ONLINE EXCLUSIVE PRICE
    434.00 €
    416.23 € / Each
    Save 17.77 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Fas (CD95; APO-1) is a 45kDa cell surface protein that mediates apoptosis when cross-linked with agonistic anti-Fas antibodies or by Fas ligand (FasL; CD178). Fas belongs to the TNF (Tumor Necrosis Factor)/NGF (Nerve Growth Factor) receptor family, and is expressed in various tissues and cells including the thymus, liver, ovary and lung. CD178 (FasL), a member of the TNF cytokine family, induces apoptosis by binding to Fas, its cell-surface receptor. FasL may exist as either membrane bound or soluble forms and is expressed by activated T and NK cells. FasL may also be constitutively expressed in some immunologically privileged sites, e.g., eye and testis. Fas and FasL play an important role in the induction of apoptosis, and thus regulate a variety of immunological responses. The NOK-1 antibody clone has been reported to recognize human FasL, recognizing both the membrane bound (FasL) and soluble (sFasL) forms. FasL and sFasL have been reported to migrate at reduced molecular weights of 40 and 26kDa, respectively. However, the molecular weights observed in a particular sample may vary according to FasL and sFasL glycosylation and breakdown patterns as described in the literature. The NOK-1 antibody clone is not
recommended for the western blot application.

Host Species: Mouse
Clone: NOK-1
Isotype: IgG1
Species Reactivity: Human
Immunogen: Mouse T lymphoma cells (L5178Y) expressing human FasL

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD178
Monoclonal
0.5mg/mL
Fas ligand; FASL; FASLG; CD95 ligand; CD95L; CD95-L; TNFSF6; APTL; APT1LG1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
NOK-1
Biotin
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse T lymphoma cells (L5178Y) expressing human FasL
0.1 mg
Immunology
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.