missing translation for 'onlineSavingsMsg'
Learn More

CD184 Rat anti-Mouse, FITC, Clone: 2B11/CXCR4, BD

Rat Monoclonal Antibody

166.92 € - 391.62 €

Specifications

Antigen CD184
Clone 2B11/CXCR4
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15801289
View Documents
BD Biosciences
561735
25 μg
ONLINE EXCLUSIVE PRICE
174.00 €
166.92 € / Each
Save 7.08 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15816748
View Documents
BD Biosciences
551967
0.1 mg
ONLINE EXCLUSIVE PRICE
408.00 €
391.62 € / Each
Save 16.38 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody 2B11/CXCR4 reacts with the mouse CD184, which is also known as CXC chemokine receptor, CXCR4. CXCR4 (previously known as fusin, LESTR and HUMSTR), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokines, SDF-1/CXCL12. Mouse CXCR4 showed 91% homology at amino acid level with human CXCR4. The CXCR4 is widely expressed by hematopoietic and non hematopoietic cell types including PMN, monocytes, T cells, B cells, CD34+ progenitor cells, endothelial cells, neurons and astrocytes. The human CXCR4 is used by T-tropic HIV-1 as a co-receptor for viral entry. The mouse CXCR4 gene has been mapped to chromosome 1. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 2B11/CXCR4
Isotype: IgG2b κ
Species Reactivity: Mouse
Immunogen: GST-NCXCR4 fusion protein

Flow Cytometry

Specifications

Specifications

CD184
0.5mg/mL
Monoclonal
Rat
Mouse
GST-NCXCR4 fusion protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
2B11/CXCR4
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2b κ
Affinity Purified
CXCR4, C-X-C chemokine receptor type 4; Fusin; LESTR; PB-CKR; Sdf1r
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.