missing translation for 'onlineSavingsMsg'
Learn More

CD2 Mouse anti-Human, BUV395, Clone: RPA-2.10, BD

Mouse Monoclonal Antibody

230.05 € - 477.65 €

Specifications

Antigen CD2
Clone RPA-2.10
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 395
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15813129
View Documents
BD Biosciences
563820
25 Tests
ONLINE EXCLUSIVE PRICE
240.00 €
230.05 € / Each
Save 9.95 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15803129
View Documents
BD Biosciences
563819
100 Tests
ONLINE EXCLUSIVE PRICE
498.00 €
477.65 € / Each
Save 20.35 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.

The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications

Specifications

CD2
Flow Cytometry
Brilliant Ultraviolet 395
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
LFA-2; Lymphocyte function antigen-2; T11/Leu-5; Rosette receptor
RPA-2.10
Monoclonal
Mouse
Human
Human Phytohemagglutinin-treated Lymphoblast
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.