Learn More
CD27 Mouse anti-Human, APC-H7, Clone: M-T271, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560223
Description
Reacts with a disulphide-linked 110kDa dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. CD27 is highly induced on T cells after TCR stimulation. CD27 may be involved in cellular interaction of T and B lymphocytes.
Host Species: Mouse
Clone: M-T271
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD27 | |
| Monoclonal | |
| APC-H7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| M-T271 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human T-CLL cells | |
| 25 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.