Learn More
CD27 Mouse anti-Human, APC-H7, Clone: M-T271, BD
Mouse Monoclonal Antibody
141.00 € - 299.00 €
Specifications
| Antigen | CD27 |
|---|---|
| Clone | M-T271 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-H7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840029
|
BD Biosciences
560223 |
25 Tests |
141.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15830029
|
BD Biosciences
560222 |
100 Tests |
299.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with a disulphide-linked 110kDa dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. CD27 is highly induced on T cells after TCR stimulation. CD27 may be involved in cellular interaction of T and B lymphocytes.
Host Species: Mouse
Clone: M-T271
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD27 | |
| Flow Cytometry | |
| APC-H7 | |
| RUO | |
| Human | |
| Human T-CLL cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| M-T271 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.